9-Fluoreneacetic acid ≥99.5%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A18772.03
A18772.06
A18772.03EA
982
DKK
A18772.03
A18772.06
9-Fluoreneacetic acid ≥99.5%
9-Fluoreneacetic acid
Purity: 99.5+%
CAS No.: 6284-80-6
Molecular Formula: C15H12O2
Formula Weight: 224.26
Melting Point: 133-135°
MDL No.: MFCD00013262
Formel:
C₁₅H₁₂O₂ Smeltepunkt: 133…135 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013262 CAS-nummer: 6284-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...