Ethyl (4-nitrobenzoyl)acetate 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A17603.09 A17603.18 A17603.30
A17603.09EA 589 DKK
A17603.09 A17603.18 A17603.30
Ethyl (4-nitrobenzoyl)acetate 97%
Ethyl (4-nitrobenzoyl)acetate

Purity: 97% CAS No.: 838-57-3 Molecular Formula: C11H11NO5 Formula Weight: 237.21 Melting Point: 68-73° Beilstein Reference: 1124763 MDL No.: MFCD00007357

Formel: C₁₁H₁₁NO₅
Smeltepunkt: 68…73 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007357
CAS-nummer: 838-57-3
EINECS: 212-656-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR