N-tert-Butylisopropylamine 98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98%
CAS No.: 7515-80-2
Molecular Formula: (CH3)3CNHCH(CH3)2
Formula Weight: 115.22
Flash Point: -3°(26°F)
Refractive Index: 1.3980
Boiling Point: 98°
Beilstein Reference: 1846752
UN No.: UN2733
MDL No.: MFCD00037059
Formel:
(CH₃)₃CNHCH(CH₃)₂ Kogepunkt: 98 °C Massefylde: 0,727 Flammepunkt: –3 °C (26 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00037059 CAS-nummer: 7515-80-2 EINECS: 231-370-3 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...