Nile blue A
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Nile blue A sulphate, Nile blue sulphate, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulphate, Basic Blue 12
A17174.14 A17174.06
A17174.14EA 951 DKK
A17174.14 A17174.06
Nile blue A
Nile blue A
CAS No.: 3625-57-8 Molecular Formula: C40H40N6O6S Formula Weight: 732.84 Melting Point: 300° dec. MDL No.: MFCD00064529
Formel: C₄₀H₄₀N₆O₆S
Molekylvægt: 732.84 g/mol
Smeltepunkt: >300 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064529
CAS-nummer: 3625-57-8
EINECS: 222-832-5