1-Cyclohexyl-2-pyrrolidone 99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 99%
CAS No.: 6837-24-7
Molecular Formula: C10H17NO
Formula Weight: 167.25
Flash Point: 140°(284°F)
Refractive Index: 1.4990
Melting Point: 12-15°
Boiling Point: 290-292°
Beilstein Reference: 121832
UN No.: UN2810
MDL No.: MFCD00003191
Formel:
C₁₀H₁₇NO Kogepunkt: 290…292 °C Smeltepunkt: 12…15 °C Massefylde: 1,007 Flammepunkt: 140 °C (284 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003191 CAS-nummer: 6837-24-7 EINECS: 229-919-7 ADR: 6.1,II |