Brucine dihydrate 98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
A16251.09 A16251.18
A16251.09EA 380 DKK
A16251.09 A16251.18
Brucine dihydrate 98%
Brucine dihydrate
Brucine dihydrate is used as a resolving agent for chiral acids.
Purity: 98% CAS No.: 5892-11-5 Molecular Formula: C23H26N2O4.2H2O Formula Weight: 430.50 (394.47anhy) Melting Point: 176-178° UN No.: UN1570 MDL No.: MFCD00149384
Formel: C₂₃H₂₆N₂O₄·2H₂O
Molekylvægt: 430.50 g/mol (394.47 anhy)
Smeltepunkt: 176…178 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149384
CAS-nummer: 5892-11-5
UN nummer: 1570
ADR: 6.1 I