Copper(II) oxalate hemihydrate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Cupric oxalate hemihydrate

A15365.18 A15365.30
A15365.18EA 492 DKK
A15365.18 A15365.30
Copper(II) oxalate hemihydrate 98%
Copper(II) oxalate hemihydrate

Purity: 98% CAS No.: 5893-66-3 Molecular Formula: CuC2O4.0.5H2O Formula Weight: 160.57 (151.51anhy) Solubility: Soluble in NH4OH UN No.: UN3288 MDL No.: MFCD00150451

Formel: CuC₂O₄·½H₂O
Molekylvægt: 160.57 g/mol (151.51 anhy)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00150451
CAS-nummer: 5893-66-3
EINECS: 212-411-4
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR