N,N-Dimethylmethyleneammonium iodide 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N,N-Dimethylmethyleneiminium Iodide, Eschenmoser's Salt
A15146.09
A15146.18
A15146.30
A15146.09EA
1100
DKK
A15146.09
A15146.18
A15146.30
N,N-Dimethylmethyleneammonium iodide 97%
N,N-Dimethylmethyleneammonium iodide
Purity: 97%
CAS No.: 33797-51-2
Molecular Formula: C3H8IN
Formula Weight: 185.01
Storage Temperature: 2°C to 8°C
Melting Point: ca 240° dec.
Beilstein Reference: 3594966
MDL No.: MFCD00011810
Formel:
C₃H₈IN Smeltepunkt: ∼ 240 °C (dec.) |
MDL Number:
MFCD00011810 CAS-nummer: 33797-51-2 EINECS: 251-680-2 Merck Index: 14,3251 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...