Sodium-2-bromoethanesulphonate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Sodium 2-bromoethanesulphonate, 2-Bromoethanesulphonic Acid Sodium Salt

A15076.14 A15076.22 A15076.36
A15076.14EA 348 DKK
A15076.14 A15076.22 A15076.36
Sodium-2-bromoethanesulphonate 98%
Sodium-2-bromoethanesulphonate

Purity: 98% CAS No.: 4263-52-9 Molecular Formula: BrCH2CH2SO3Na Formula Weight: 211.02 Melting Point: ca 284° dec. Beilstein Reference: 4590828 MDL No.: MFCD00007530

Formel: C₂H₅BrO₃S
Smeltepunkt: ∼ 284 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007530
CAS-nummer: 4263-52-9
EINECS: 224-244-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR