2-Phenyl-2-pentenal mixture E/Z ≥90%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A13635.06 A13635.14
A13635.06EA 503 DKK
A13635.06 A13635.14
2-Phenyl-2-pentenal mixture E/Z ≥90%
2-Phenyl-2-pentenal

Purity: 90+% CAS No.: 3491-63-2 Molecular Formula: C11H12O Formula Weight: 160.22 Storage Temperature: 2°C to 8°C Flash Point: 100°(212°F) Refractive Index: 1.5510 Boiling Point: 124-126°/14mm Beilstein Reference: 1859032 MDL No.: MFCD00051835

Formel: C₁₁H₁₂O
Kogepunkt: 124…126 °C (14 mmHg)
Massefylde: 1,005
Flammepunkt: >100 °C (212 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00051835
CAS-nummer: 3491-63-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR