3-Isocyanatopropyltriethoxysilane 95%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 95%
CAS No.: 24801-88-5
Molecular Formula: C10H21NO4Si
Formula Weight: 247.37
Flash Point: 77°(170°F)
Refractive Index: 1.4210
Odour: Mild
Boiling Point: 238°
Beilstein Reference: 4404652
UN No.: UN2927
MDL No.: MFCD00051459
Formel:
C₁₀H₂₁NO₄Si Kogepunkt: 238 °C Massefylde: 0,990 Flammepunkt: 77 °C (170 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00051459 CAS-nummer: 24801-88-5 EINECS: 246-467-6 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...