Ethyl hydrogen malonate 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A12627.09 A12627.18 A12627.30
A12627.09EA 567 DKK
A12627.09 A12627.18 A12627.30
Ethyl hydrogen malonate 96%
Ethyl hydrogen malonate

Purity: 96% CAS No.: 1071-46-1 Molecular Formula: C5H8O4 Formula Weight: 132.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.119 Refractive Index: 1.4295 Appearance: Colourless to yellow Boiling Point: 106°/3mm Beilstein Reference: 1758845 MDL No.: MFCD00020490

Formel: C₅H₈O₄
Kogepunkt: 106 °C (3 mmHg)
Massefylde: 1,119
Flammepunkt: >110 °C (230 °F)
MDL Number: MFCD00020490
CAS-nummer: 1071-46-1
EINECS: 213-992-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR