Titanium(IV) 2-ethylhexyloxide ≥95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrakis(2-ethylhexyl) Orthotitanate, Titanium(IV) 2-Ethylhexyloxide, Tetrakis(2-ethylhexyl)orthotitanate(IV)

77107.22 77107.36
77107.22EA 344 DKK
77107.22 77107.36
Titanium(IV) 2-ethylhexyloxide ≥95%
Titanium(IV) 2-ethylhexyloxide

Purity: 95% CAS No.: 1070-10-6 Molecular Formula: C32H68O4Ti Formula Weight: 564.79 Flash Point: 71°(160°F) Physical Form: Liquid Density: 0.926 Refractive Index: 1.482 Sensitivity: Moisture Sensitive Appearance: Pale yellow Melting Point: -25° Boiling Point: 194°/0.25mm Beilstein Reference: 3692023 MDL No.: MFCD00015264

Formel: C₃₂H₆₈O₄Ti
Molekylvægt: 564.79 g/mol
Kogepunkt: 194 °C (0.25 mmHg)
Smeltepunkt: < –25 °C
Massefylde: 0.926
Flammepunkt: 71 °C (160 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015264
CAS-nummer: 1070-10-6
EINECS: 213-969-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR