Holmiumtrichlorid, ultra dry ≥99.95% (metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Holmium trichloride
Ampuled under argon
47178.06
47178.14
47178.06EA
2820
DKK
47178.06
47178.14
Holmiumtrichlorid, ultra dry ≥99.95% (metals basis)
Holmiumtrichlorid
Purity: 99.95%
CAS No.: 10138-62-2
Molecular Formula: HoCl3
Formula Weight: 271.29
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.70
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 718°
MDL No.: MFCD00011051
Formel:
Cl₃Ho Smeltepunkt: 718 °C Massefylde: 3,70 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011051 CAS-nummer: 10138-62-2 EINECS: 233-387-1 Merck Index: 14,4727 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...