Aluminiumiodid, ultra dry ≥99.999% (metals basis)
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Aluminum iodide
Ampuled under argon
Purity: 99.999%
CAS No.: 7784-23-8
Molecular Formula: AlI3
Formula Weight: 407.69
Physical Form: Powder, Ampouled under argon
Density: 3.98
Sensitivity: Moisture and Light Sensitive
Appearance: White to brownish-purple
Odour: Pungent
Melting Point: 191°
Boiling Point: 360°
UN No.: UN3260
MDL No.: MFCD00003427
Formel:
AlI₃ Molekylvægt: 407.7 g/mol Kogepunkt: 360 °C Smeltepunkt: 191 °C Massefylde: 3.98 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003427 CAS-nummer: 7784-23-8 EINECS: 232-054-8 UN nummer: 3260 ADR: 8 II Merck Index: 14,346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...