Phosphorus, plasma standard solution, P 1,0000 µg/ml, Specpure®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
47152.AE
47152.AP
47152.AEEA
1800
DKK
47152.AE
47152.AP
Phosphorus, plasma standard solution, P 1,0000 µg/ml, Specpure®
Standarder
Kationiske standarder
Phosphorus Standards
Molecular Formula: H₃PO₄, 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00133771