Dysprosiumtribromid, ultra dry ≥99.99% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Ampuled under argon

47127.03 47127.06
47127.03EA 1370 DKK
47127.03 47127.06
Dysprosiumtribromid, ultra dry ≥99.99% (metals basis)
Dysprosiumtribromid

Purity: 99.99% CAS No.: 14456-48-5 Molecular Formula: DyBr3 Formula Weight: 402.23 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Hygroscopic Melting Point: 881° Boiling Point: 1480° MDL No.: MFCD00016069

Formel: Br₃Dy
Molekylvægt: 402.21 g/mol
Kogepunkt: 1480 °C
Smeltepunkt: 881 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016069
CAS-nummer: 14456-48-5
EINECS: 238-443-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR