3,3'-Diallylbisphenol A diacetate epoxy curative EC-392
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
46403.14
46403.22
46403.14EA
768
DKK
46403.14
46403.22
3,3'-Diallylbisphenol A diacetate epoxy curative EC-392
3,3'-Diallylbisphenol A diacetate
CAS No.: 1071466-61-9
Molecular Formula: C25H28O4
Formula Weight: 392.5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Amber
MDL No.: MFCD22200539
Applications: Recommended for use in epoxy systems or in combination with acrylates, methacrylates, and maleimides
Formel:
C₂₅H₂₈O₄ Molekylvægt: 392.5 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD22200539 CAS-nummer: 1071466-61-9 |