Zeolite ZSM-5, ammonium
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
45880.14
45880.22
45880.36
45880.14EA
592
DKK
45880.14
45880.22
45880.36
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 400 m2/g, 30:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
Formel:
SiO₂:Al₂O₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00132601 CAS-nummer: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...