Zeolite ZSM-5, ammonium
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
45879.14
45879.22
45879.36
45879.14EA
605
DKK
45879.14
45879.22
45879.36
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 425 m2/g, 23:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
Formel:
SiO₂:Al₂O₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00132601 CAS-nummer: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...