Palladium 5% on activated carbon paste (50% water) A405032-5
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
45499.06 45499.14 45499.22
45499.06EA 1070 DKK
45499.06 45499.14 45499.22
Palladium 5% on activated carbon paste (50% water) A405032-5
Palladium
CAS No.: 7440-05-3 Appearance: Black Odour: Odourless MDL No.: MFCD03457879
Formel: Pd
Molekylvægt: 106.420 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011167
CAS-nummer: 7440-05-3
EINECS: 231-115-6