Palladium 5% on activated carbon paste (50% water) 5R58
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
45132.22 45132.14 45132.06
45132.22EA 20110 DKK
45132.22 45132.14 45132.06
Palladium 5% on activated carbon paste (50% water) 5R58
Palladium
CAS No.: 7440-05-3 Appearance: Black Odour: Odourless MDL No.: MFCD03457879
Formel: Pd
Molekylvægt: 106.420 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03457879
CAS-nummer: 7440-05-3
EINECS: 231-115-6