Aluminum oxide-Silicon oxide 13%, low surface area macroporous catalyst support
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43863.A1
43863.A7
43863.A1EA
3100
DKK
43863.A1
43863.A7
Aluminum oxide-Silicon oxide 13%, low surface area macroporous catalyst support
Aluminum oxide-Silicon oxide
Physical Form: 3/16" Spheres
MDL No.: MFCD00058866