Mesoporphyrin IX dihydrochloride ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43320.MD
43320.MF
43320.MDEA
5430
DKK
43320.MD
43320.MF
Mesoporphyrin IX dihydrochloride ≥97%
Mesoporphyrin IX dihydrochloride
Purity: 97%
CAS No.: 68938-72-7
Molecular Formula: C24H38N4O4.2HCl
Formula Weight: 639.62
Physical Form: Crystalline
MDL No.: MFCD00136049
Formel:
C₂₄H₃₈N₄O₄.2HCl Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00136049 CAS-nummer: 68938-72-7 EINECS: 273-141-0 |