Polyethyleneimine, on silica beads 20-40 mesh anion exchange resin
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43302.AB
43302.AK
43302.AD
43302.ABEA
487
DKK
43302.AB
43302.AK
43302.AD
Polyethyleneimine, on silica beads 20-40 mesh anion exchange resin
Polyethyleneimine
CAS No.: 9002-98-6
Physical Form: Beads
Sensitivity: Hygroscopic
MDL No.: MFCD00084427
Applications: For the separation of nucleotide mixtures
Formel:
(CH2CH2NH)n Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00084427 CAS-nummer: 9002-98-6 |