Tetraallylsilane ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
|
43183.03
43183.06
43183.03EA
592
DKK
43183.03
43183.06
Tetraallylsilane ≥97%
Tetraallylsilane
Purity: 97%
CAS No.: 1112-66-9
Molecular Formula: (H2C=CHCH2)4Si
Formula Weight: 192.38
Flash Point: 77°(171°F)
Physical Form: Liquid
Density: 0.8314
Refractive Index: 1.4850
Boiling Point: 91°/10mm
MDL No.: MFCD00048161
Applications: Synthesis of dendrimers
Formel:
(H₂C=CHCH₂)₄Si Kogepunkt: 91 °C (10 mmHg) Massefylde: 0,8314 Flammepunkt: 77 °C (171 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00048161 CAS-nummer: 1112-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...