Lanthanum(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43122.22
43122.36
43122.22EA
751
DKK
43122.22
43122.36
Lanthanum(III) oxalate hydrate ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) oxalate hydrate
CAS No.: 79079-18-8
Molecular Formula: C6La2O12.xH2O
Formula Weight: 541.87(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: dec.
UN No.: UN3288
MDL No.: MFCD00150540
Formel:
C₆La₂O₁₂·xH₂O Molekylvægt: 541.87 g/mol (anhy) |
MDL Number:
MFCD00150540 CAS-nummer: 79079-18-8 |