Silica Gel, TLC High Purity Grade, without Binder, with Fluorescent Indicator
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42765.18 42765.30
42765.18EA 265 DKK
42765.18 42765.30
Silica Gel, TLC High Purity Grade, without Binder, with Fluorescent Indicator
Kromatografi sorbenter TLC Sorbenter
This TLC high purity grade silica gel without binder, with fluorescent indicator is used for thin layer chromatography plate preparation.
  • pH 6,5 to 7,5