Silica gel, TLC high purity grade, with gypsum binder
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42763.18 42763.30
42763.18EA 469 DKK
42763.18 42763.30
Silica gel, TLC high purity grade, with gypsum binder
Kromatografi sorbenter TLC Sorbenter
This TLC high purity grade silica gel with gypsum binder is used for thin layer chromatography plate preparation.
  • pH 6 to 7