Polyethylene oxide, M.W. 300,000
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
CAS No.: 25322-68-3
Molecular Formula: (-CH2CH2O-)n
Physical Form: Powder
Density: 1.21
Refractive Index: 1.4539
Appearance: White to off-white
Odour: Odourless
pH: 5-7
Melting Point: 65°
MDL No.: MFCD00147979
Formel:
(-CH₂CH₂O-)n Smeltepunkt: 65 °C Massefylde: 1,21 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00147970 CAS-nummer: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...