Silver, brazing alloy (Ag:Sn; 3.5:96.5 wt%), Ø 3.0 mm (0.13 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41030.18
41030.30
41030.TD
41030.18EA
616
DKK
41030.18
41030.30
41030.TD
Silver, brazing alloy (Ag:Sn; 3.5:96.5 wt%), Ø 3.0 mm (0.13 in)
Silver, brazing alloy
Molecular Formula: Ag:Sn; 3.5:96.5 wt%
Physical Form: Nontoxic, cadmium and lead free
MDL No.: MFCD00801108