Zirkon(IV)oxid 18% in water stabilised, colloidal dispersion
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Zirconium dioxide, Zirconium Oxide
Stabilization: 1.3% Stabilised with yttria (Y₂O₃)
40123.36 40123.A4
40123.36EA 1370 DKK
40123.36 40123.A4
Zirkon(IV)oxid 18% in water stabilised, colloidal dispersion
Zirkon(IV)oxid
stabilized with 1.3% yttrium oxide
CAS No.: 1314-23-4 Molecular Formula: ZrO2/Y2O3 Physical Form: 0.1 Micron Particles in Liquid MDL No.: MFCD00011310
Applications: In ceramic industry
Formel: O₂Zr
Molekylvægt: 123.22 g/mol
Kogepunkt: 4300
Smeltepunkt: 2680
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011310
CAS-nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180