Tin(II) tartrate hydrate ≥95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

39604.14 39604.22
39604.14EA 2010 DKK
39604.14 39604.22
Tin(II) tartrate hydrate ≥95%
Tin(II) tartrate hydrate

Purity: 95% CAS No.: 815-85-0 Molecular Formula: C4H4O6Sn.xH2O Formula Weight: 266.76(anhy) Physical Form: Powder Sensitivity: Air Sensitive Odour: Odourless MDL No.: MFCD00054358

Formel: C₄H₄O₆Sn·xH₂O
Molekylvægt: 266.78 g/mol (anhy)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00054358
CAS-nummer: 815-85-0
EINECS: 212-427-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR