Cesium titanium oxide ≥99.85% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

39207.06 39207.14
39207.06EA 775 DKK
39207.06 39207.14
Cesium titanium oxide ≥99.85% (metals basis)
Cesium titanium oxide

Purity: 99.85% CAS No.: 12158-57-5 Molecular Formula: Cs2O3Ti Formula Weight: 361.71 Physical Form: -100 Mesh Powder Appearance: White MDL No.: MFCD00053969

Formel: Cs₂O₃Ti
Molekylvægt: 361.71 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00053969
CAS-nummer: 12158-57-5
EINECS: 235-283-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR