Fluoride, Ion chromatography standard solution, Specpure®, F- 1000µg/ml
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
35550.AE 35550.AP
35550.AEEA 1800 DKK
35550.AE 35550.AP
Fluoride, Ion chromatography standard solution, Specpure®, F- 1000µg/ml
Standarder Anioniske standarder Fluoride Standards
NaF in H₂O; MDL No.: MFCD00144315
Bestil nu
Specifikationer
Environmentally Preferable
MDL No. MFCD00144315
Molecular formula NaF in H2O
Physical form Liquid