Boats, ceramic, AL23
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
33171.KF 33172.KF 33173.KF 33174.KF 33176.KF 33179.KF 33180.KF 33181.KF
33171.KFEA 787 DKK
33171.KF 33172.KF 33173.KF 33174.KF 33176.KF 33179.KF 33180.KF 33181.KF
Boats, ceramic, AL23
Vejebåde Forbrændingsbåde
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.