Ceramic plates, rectangular or square, AL25
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
33159.KF 33160.KF
33159.KFEA 2170 DKK
33159.KF 33160.KF
Ceramic plates, rectangular or square, AL25
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
Pure Al₂O₃ Aluminium oxide, AL25.
Very porous, good thermal insulation, highest resistance to thermal shock of all the Al₂O₃ materials