Ceramic tubes, both ends open, thin wall
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31706.DR 31706.DM 31708.DM 31708.DR 31708.DW 31709.DM 31709.DR 31709.DW
31706.DREA 760 DKK
31706.DR 31706.DM 31708.DM 31708.DR 31708.DW 31709.DM 31709.DR 31709.DW
Ceramic tubes, both ends open, thin wall
Slanger Overførselsslange
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.