Nikkel on silica-alumina for use as a catalyst
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
31276.06 31276.14 31276.22
31276.06EA 276 DKK
31276.06 31276.14 31276.22
Nikkel on silica-alumina for use as a catalyst
Nikkel
CAS No.: 7440-02-0 Molecular Formula: 66+/-5% Ni Physical Form: Powder Appearance: Grey Odour: Odourless UN No.: UN3178 MDL No.: MFCD00011137
Formel: Ni
Molekylvægt: 58.69 g/mol
Kogepunkt: 3007
Smeltepunkt: 1453
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011137
CAS-nummer: 7440-02-0
UN nummer: 3178
ADR: 4.1 II
Merck Index: 12,06582