Zirconium silicate
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
30477.A3
30477.A9
30477.36
30477.A3EA
1110
DKK
30477.A3
30477.A9
30477.36
Zirconium silicate
Zirconium silicate
CAS No.: 10101-52-7
Molecular Formula: ZrSiO4
Formula Weight: 183.30
Physical Form:
Formel:
O₄SiZr Molekylvægt: 183.305 g/mol Massefylde: 4.56 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00085353 CAS-nummer: 10101-52-7 EINECS: 233-252-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...