Europium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

13212.03 13212.06 13212.14
13212.03EA 876 DKK
13212.03 13212.06 13212.14
Europium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Europium(III) carbonate hydrate

Purity: 99.99% CAS No.: 86546-99-8 Molecular Formula: Eu2(CO3)3.xH2O Formula Weight: 483.95(anhy) Physical Form: Powder Odour: Odourless MDL No.: MFCD00149702

Formel: Eu₂(CO₃)₃·xH₂O
Molekylvægt: 483.95 g/mol (anhy)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149702
CAS-nummer: 86546-99-8
EINECS: 227-582-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR