Yttrium cobalt 99,9 REO basis, lumps, REacton®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
12952.14
12952.22
12952.14EA
1630
DKK
12952.14
12952.22
Yttrium cobalt 99,9 REO basis, lumps, REacton®
Yttrium cobalt
Purity: 99.9%
Molecular Formula: Y:Co; 84:15 wt% (Ca 1%)
MDL No.: MFCD00209676
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...