Ytterbium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Ytterbium trinitrate hydrate
12901.09
12901.18
12901.09EA
1060
DKK
12901.09
12901.18
Ytterbium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Ytterbium(III) nitrate hydrate
Purity: 99.9%
Molecular Formula: Yb(NO3)3.xH2O
Formula Weight: 359.05(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
UN No.: UN1477
MDL No.: MFCD00149883
Formel:
Yb(NO₃)₃·xH₂O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149883 EINECS: 237-384-6 UN nummer: 1477 ADR: 5.1,III Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...