Aluminiumoxid 20% in water, colloidal dispersion
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Aluminium(III) oxide, Alumina C, Aluminum oxide
12733.A1 12733.4Z
12733.A1EA 613 DKK
12733.A1 12733.4Z
Aluminiumoxid 20% in water, colloidal dispersion
Aluminium oxide
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 0.05 Micron Particles Odour: Odourless MDL No.: MFCD00003424
Applications: Effective in high temperature refractories as a stable binder
Formel: Al₂O₃
Molekylvægt: 101.96 g/mol
Kogepunkt: 2980
Smeltepunkt: 2045
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003424
CAS-nummer: 1344-28-1
Merck Index: 13,00355