Strontium zirconium oxide ≥99.3% (metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Strontium zirconate
12346.30
12346.A1
12346.30EA
676
DKK
12346.30
12346.A1
Strontium zirconium oxide ≥99.3% (metals basis)
Strontium zirconium oxide
Purity: 99.3%
CAS No.: 12036-39-4
Molecular Formula: SrZrO3
Formula Weight: 226.84
Physical Form: -325 Mesh Powder
Density: 5.5
Appearance: White to off-white
Odour: Odourless
Melting Point: 2700°
Boiling Point: 9000°
MDL No.: MFCD00049555
Formel:
SrZrO₃ Kogepunkt: 9000 °C Smeltepunkt: 2700 °C Massefylde: 5,5 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00049555 CAS-nummer: 12036-39-4 EINECS: 234-847-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...