Wastewater inorganics quality control standard
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2833249 2833149
HACH2833249EA 1410 DKK
HACH2833249 HACH2833149
Wastewater inorganics quality control standard
Standarder Miljø analyse standarder
These mixed standards are ready to use, no dilutions are required. They contain ammonia nitrogen, nitrate nitrogen, COD, phosphate, sulphate and TOC.