Storage vials
Leverandør: SAVILLEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
200-003-20 200-007-20 200-015-20 200-030-12
SAVI200-003-20EA 157 DKK
SAVI200-003-20 SAVI200-007-20 SAVI200-015-20 SAVI200-030-12
Storage vials
Hætteglas
PFA vials with round or conical interior.
  • Ideal for storage and digestion in geochemistry
  • Flat base for maximum heating efficiency
  • Square exterior vials with hex closures have higher pressure rating
Bestillingsoplysninger: Closures can be ordered separately. Modifications may be available on request.