Testkits, chlortest, til Spectroquant®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.00597.0001 1.00598.0001 1.00598.0002 1.00599.0001 1.00602.0001 1.00602.0002 1.00595.0001 1.00086.0001 1.00087.0001 1.00088.0001
1.00597.0001EA 852 DKK
1.00597.0001 1.00598.0001 1.00598.0002 1.00599.0001 1.00602.0001 1.00602.0002 1.00595.0001 1.00086.0001 1.00087.0001 1.00088.0001
Testkits, chlortest, til Spectroquant®, Supelco®
Miljø test kit
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*4 Reagenser til analyse af frit chlor er reagens Cl₂-1 1.00086.0001 og reagens Cl₂-2 1.00087.0001 sammen med tom kuvette og tilbehør 1.00089.0001
Reagenser til total chlor er reagens Cl₂-1 1.00086.0001, reagens Cl₂-2 1.00087.0001 og reagens Cl₂-3 1.00088.0001 sammen med tom kuvette og tilbehør 1.00089.0001
*7 Denne metode er officielt anerkendt af USEPA som en alternativ metode til undersøgelse af følgende:
*7-1 spildevand
*7-2 drikkevand
*7-3 hhv. drikke- og spildevand