Industrial gloves, uvex phynomic lite w
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6004106 6004107 6004108 6004109 6004110 6004111
112-4930EA 45.1 DKK
112-4930 112-4931 112-4932 112-4933 112-4934 112-4935
Industrial gloves, uvex phynomic lite w
Handsker
The uvex phynomic lite w is the lightest safety glove in its class. Palm and fingertips feature a breathable aqua-polymer impregnation which is extremely thin, but still highly durable.
  • Exceptional dexterity
  • Excellent dry grip
  • Very high level of breathability
Ideal for precision assembly work and precision work. Suitable for dry applications.
Certificeringer: EN 388 (2121X)
Food contact approved
OEKO-TEX® Standard 100.