Industrial gloves, uvex phynomic allround
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6004906 6004907 6004908 6004909 6004910 6004911
112-4936EA 44.3 DKK
112-4936 112-4937 112-4938 112-4939 112-4940 112-4941
Industrial gloves, uvex phynomic allround
Handsker
The uvex phynomic is a lightweight safety glove which is suitable for a wide range of applications. The polyamide/elastane glove liners feature an aqua-polymer foam coating on palm and fingertips.
  • Foam coating is extremely flexible, provides good grip and leaves no trace on sensitive surfaces
  • Dark colour makes the gloves particularly dirt resistant
  • Excellent dexterity and grip
  • Very high level of breathability
Ideal for assembly and precision work. Suitable for use in dry and slightly damp areas.
Certificeringer: EN 388 (3131X)
OEKO-Tex® Standard 100